Order Entry
Austria
ContactUsLinkComponent
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP) 5 mg, tablets
Catalog # J61401.KN
CAS Number:  
4-Nitrophenyl phosphate disodium salt hexahydrate (pNPP) 5 mg, tablets
Catalog # J61401.KN
CAS Number:  

Specifications

  • Pk:
    100
  • Seq:
    0100

Specifications

About this item

 :
Melting Pt:300 °C
 :371.144 g/mol
Storage Temperature:Refrigerator
EINECS:224-246-5
CAS Number:333338-18-4
 :71735-29-0
Formula:C₆H₄NNa₂O₆P·6H₂O
MDL Number:MFCD00066288