5-Hydroxyindol-3-ylacetic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:54-16-0
:
Formula:C₁₀H₉NO₃
MDL Number:MFCD00005639
:191.19 g/mol
Melting Pt:161…164 °C (dec.)
Storage Temperature:Freezer
EINECS:200-195-4
Related Information
Frequently Bought Together