Order Entry
Austria
ContactUsLinkComponent
Sodium chloride 5 M in aqueous solution, autoclaved
Sodium chloride 5 M in aqueous solution, autoclaved
Catalog # J61890.AK
CAS Number:  
Sodium chloride 5 M in aqueous solution, autoclaved
Catalog # J61890.AK
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Saline, NaCl, Sea salt, Halite
 :
 :58.44 g/mol
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477