


Trichloroacetic acid (TCA) 5% (w/v) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:76-03-9
:
Formula:Cl₃CCOOH
MDL Number:MFCD00004177
:163.39 g/mol
Merck Index:14,9627
Storage Temperature:Ambient
EINECS:200-927-2
UN:2564
ADR:8,III
:GHS05
:GHS07
:GHS09
Related Information
Frequently Bought Together