Sodium pyruvate cell culture reagent
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:113-24-6
:
Formula:CH₃COCOONa
MDL Number:MFCD00002586
:110.04 g/mol
Melting Pt:220…230 °C
Density:1.718 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-024-4
Related Information
Frequently Bought Together