Order Entry
Austria
ContactUsLinkComponent
 
L(+)-Valine 98.5-101.5% (dried basis) USP, Multi-Compendial, J.T.Baker®
Catalog # 2095-06
Supplier:  Avantor
CAS Number:  
undefined
L(+)-Valine 98.5-101.5% (dried basis) USP, Multi-Compendial, J.T.Baker®
Catalog # 2095-06
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    1 kg
  • Seq:
    4024
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Meets E.P. Chemical Specifications:
    98.5 - 101.5%
  • GMP Manufactured Product:
    Passes Test
  • Meets U.S.P Requirements:
    26.6 - 28.8 Degree
  • Meets J.P. Chemical Specifications:
    5.5 - 7.0
  • Meets B.P. Chemical Specifications:
    ≤0.3%
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤0.1%
  • Bulk Pharmaceutical Chemical:
    ≤0.05%
  • USP - Assay (C₅H₁₁NO₂) (dried basis):
    98.5 - 101.5 %
  • USP - Identification:
    Passes Test
  • USP - Specific Rotation [Å]²⁵ ^D (+):
    26.6 - 28.8 Degree
  • USP - pH:
    5.5 - 7.0
  • USP - Loss on Drying at 105°C:
    ≤0.3 %
  • USP - Residue on Ignition:
    ≤0.1 %
  • USP - Chloride (Cl):
    ≤0.05 %
  • USP - Sulfate (SO₄):
    ≤0.03 %
  • USP - Iron (Fe):
    ≤0.003 %
  • USP - Related Compounds - Individual Impurities:
    ≤0.5 %
  • USP - Related Compounds - Total Impurities:
    ≤2.0 %
  • EP/BP - Assay (C₅H₁₁NO₂) (dried basis):
    98.5 - 101.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Specific Rotation [Å]²º^D (+):
    26.5 - 29.0 Degree
  • EP/BP-Ninhydrin-Positive Substances-Impurity B:
    ≤0.4 %
  • EP/BP-Ninhydrin-Positive Substances-Each:
    ≤0.2 %
  • EP/BP-Ninhydrin-Positive Substances-Total Impurities:
    ≤1.0 %
  • EP/BP - Chloride (Cl):
    ≤200 ppm
  • EP/BP - Sulfate (SO₄):
    ≤300 ppm
  • EP/BP - Ammonium (NH₄):
    ≤0.02 %
  • EP/BP - Iron (Fe):
    ≤10 ppm
  • EP/BP - Loss on Drying:
    ≤0.5 %
  • EP/BP - Ash (sulfated):
    ≤0.1 %
  • JP - Assay (C₅H₁₁NO₂) (dried basis):
    98.5 - 101.0 %
  • JP - Identification:
    Passes Test
  • JP - Optical Rotation (+):
    26.5 - 29.0 Degree
  • JP - pH:
    5.5 - 6.5
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Chloride (Cl):
    ≤0.021 %
  • JP - Sulfate (SO₄):
    ≤0.028 %
  • JP - Ammonium (NH₄):
    ≤0.02 %
  • JP - Heavy Metals (as Pb):
    ≤20 ppm
  • JP - Arsenic (As):
    ≤2 ppm
  • JP - Related Substances:
    Passes Test
  • JP - Loss on Drying at 105°C:
    ≤0.30 %
  • JP - Residue on Ignition:
    ≤0.10 %

Specification Test Results

About this item

Synonyms: H-Val-OH, Valine, (S)-(+)-Valine
 :
 :117.15 g/mol
Merck Index:13,09458
Boiling Pt:217 °C (1013 hPa)
Melting Pt:86 °C
Density:1.284 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-773-6
CAS Number:72-18-4
Formula:C₅H₁₁NO₂
MDL Number:MFCD00064220