Order Entry
Austria
ContactUsLinkComponent
Spermidine trihydrochloride ≥99%
Irritant
Spermidine trihydrochloride ≥99%
Catalog # J61595.MC
CAS Number:  
Irritant
Spermidine trihydrochloride ≥99%
Catalog # J61595.MC
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications

About this item

 :
 :254.63 g/mol
Merck Index:14,8742
Melting Pt:250 °C (dec.)
EINECS:206-379-0
 :GHS07
Storage Temperature:Refrigerator
CAS Number:334-50-9
Formula:C₇H₂₂Cl₃N₃
MDL Number:MFCD00012918