N,N-Dimethylformamide ≥99.8% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
CAS Number:68-12-2
:
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284
:73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–61 °C
Density:0.9445 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
:Management of Change (MOC) category = N
Flash Pt:57.5 °C (closed cup)
REACH:01-2119475605-32
:GHS02
:GHS08
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:= 99.8 %
- Assay (HCON(CH₃)₂) (by GC, corrected for water):≥ 99.8 %
- Appearance:Passes Test
- Color (APHA):≤ 15
- Residue after Evaporation:≤ 0.0050 %
- Titrable Acid (meq/g):≤ 0.0005
- Titrable Base (meq/g):≤ 0.003
- Water (H₂O)(by Karl Fischer titrn):≤ 0.1 %
- Trace Impurities - Iron (Fe):≤ 0.1 ppm
Specification Test Results
Frequently Bought Together