

Acrylamide 30% in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79-06-1
:
Formula:C₃H₅NO
MDL Number:MFCD00008032
:71.08 g/mol
Merck Index:14,129
Storage Temperature:Refrigerator
EINECS:201-173-7
UN:3426
ADR:6.1,III
:GHS08
:GHS07
Related Information
Frequently Bought Together