
Dithiothreitol (DTT, Cleland's reagent) ≥99% for electrophoresis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3483-12-3
:27565-41-9
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00004877
:154.25 g/mol
Merck Index:14,3376
Boiling Pt:125…130 °C (12 mmHg)
Melting Pt:40…43 °C
Flash Pt:>110 °C (230 °F)
EINECS:222-468-7
:GHS07
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together