L(+)-Isoleucine cell culture reagent
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:73-32-5
:
Formula:H₃CCH₂CH(CH₃)CH(NH₂)COOH
MDL Number:MFCD00064222
:131.17 g/mol
Merck Index:14,5179
Melting Pt:∼ 287 °C (dec.)
Storage Temperature:Ambient
EINECS:200-798-2
Related Information
Frequently Bought Together