Order Entry
Austria
ContactUsLinkComponent
N-Methyl-2-pyrrolidone (NMP) ≥99.0%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
N-Methyl-2-pyrrolidone (NMP) ≥99.0%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # 6392-08
CAS Number:  
undefined
N-Methyl-2-pyrrolidone (NMP) ≥99.0%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # 6392-08
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Assay (C₅H₉NO):
    ≥99.0 %
  • Chloride (Cl):
    ≤1 ppm
  • Color (APHA):
    ≤50
  • Free Amines (as CH₃NH₂):
    ≤0.010 %
  • Residue after Ignition:
    ≤5 ppm
  • Water (H₂O)(by Karl Fischer titrn):
    ≤0.05 %
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 nm:
    ≤0.010
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:
    ≤0.03
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 nm:
    ≤0.20
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 285 nm:
    ≤0.40
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 275 nm:
    ≤1.00

Specification Test Results

About this item

 :
 :99.13 g/mol
Merck Index:13,06140
Boiling Pt:202 °C (1013 hPa)
Melting Pt:–24 °C
Density:1.033 g/cm³ (20 °C)
Flash Pt:91 °C
EINECS:212-828-1
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:872-50-4
Formula:C₅H₉NO
MDL Number:MFCD00003193