Potassium iodate 0.05 M KIO3 (0.3N) volumetric solution, Fluka™
Artikelnr. 34274-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
About this item
:
:214 g/mol
Density:1.005 g/cm³
Storage Temperature:Ambient
EINECS:231-831-9
CAS Number:7758-05-6
Formula:KlO₃
MDL Number:MFCD00011406