Order Entry
Austria
ContactUsLinkComponent
Potassium iodate concentrate for dilution to 1L, 1/60 M KIO3 (0.1N) volumetric solution, Fluka™
Potassium iodate concentrate for dilution to 1L, 1/60 M KIO3 (0.1N) volumetric solution, Fluka™
Catalog # 38120-1EA
CAS Number:  
Potassium iodate concentrate for dilution to 1L, 1/60 M KIO3 (0.1N) volumetric solution, Fluka™
Catalog # 38120-1EA
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
 :214 g/mol
Boiling Pt:100 °C
Storage Temperature:Ambient
EINECS:231-831-9
ADR:5.1,II
CAS Number:7758-05-6
Formula:KlO₃
MDL Number:MFCD00011406