Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)

Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)

TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer), 4 L
Catalog # 41006.
Supplier:   Biotium
Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)
Catalog # 41006.
Supplier:  Biotium
CAS Number:  
Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)

TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer), 4 L

CAS Number:  
Price
106,00 €
Log in for availability
1 * 4 L
Product Details & Documents
Specifications

Specifications

  • Pk:
    4 L
  • Description:
    TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer)

Specifications