


Tetramethylammonium hydroxide 2.38% (w/w) in aqueous solution , 99,9999% (metals basis), Electronic Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-59-2
:
Formula:C₄H₁₃NO
MDL Number:MFCD00008280
:91.15 g/mol
Merck Index:14,9224
Boiling Pt:110 °C (1013 hPa)
Density:1.02 g/cm³ (20 °C)
EINECS:200-882-9
UN:1835
ADR:8,III
:GHS06
:GHS08
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together