Acetonitrile-[1-¹³C,¹⁵N] (99% ¹³C , 98% N), Sigma-Aldrich®
Catalog # SIAL485179-250MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
ADR:3,II
:M-Clarity™ quality level = MQ200
UN:1648
CAS Number:14320-89-9
Formula:C₂H₃N
MDL Number:MFCD01075385