Naled
Catalog # EHERC15410000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
- Environmentally Preferable:
Specifications
About this item
:
EINECS:206-098-3
CAS Number:300-76-5
Formula:C₄H₇Br₂Cl₂O₄P
MDL Number:MFCD00053227