Order Entry
Austria
ContactUsLinkComponent
Potassium iodate concentrate for dilution to 1/60 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Potassium iodate concentrate for dilution to 1/60 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4676.
Supplier:  Avantor
CAS Number:  
undefined
Potassium iodate concentrate for dilution to 1/60 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4676.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass ampoule
  • Pk:
    1 Ampoul
  • Seq:
    0501

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    0.0998-0.1002
  • Normality (by titrimetry):
    Passes Test

Specification Test Results

About this item

 :
 :214 g/mol
Storage Temperature:Ambient
EINECS:231-831-9
CAS Number:7758-05-6
Formula:KlO₃
MDL Number:MFCD00011406