Order Entry
Austria
ContactUsLinkComponent
Acetonitrile-[1-¹³C] (99% ¹³C ), Sigma-Aldrich®
Catalog # SIAL485160-500MG
Supplier:  Merck
CAS Number:  
Acetonitrile-[1-¹³C] (99% ¹³C ), Sigma-Aldrich®
Catalog # SIAL485160-500MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
ADR:3,II
 :M-Clarity™ quality level = MQ200
UN:1648
CAS Number:31432-55-0
Formula:C₂H₃N