Order Entry
Austria
ContactUsLinkComponent
Herbicide reference standard, Flufenoxuron 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Herbicide reference standard, Flufenoxuron 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL13712000CY
Supplier:  EHRENSTORFER
CAS Number:  
Herbicide reference standard, Flufenoxuron 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL13712000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    101463-69-8
  • Description:
    Flufenoxuron 10 µg/ml in cyclohexane
  • Formula:
    C₂₁H₁₁ClF₆N₂O₃
  • Description:
    Flufenoxuron

Specifications

About this item