Order Entry
Austria
ContactUsLinkComponent
Acetic anhydride-[D₆] (98.5% D) for NMR spectroscopy
Acetic anhydride-[D₆] (98.5% D) for NMR spectroscopy
Catalog # ACRO174670050
CAS Number:  
undefined
Acetic anhydride-[D₆] (98.5% D) for NMR spectroscopy
Catalog # ACRO174670050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • D-Enrichment:
    ≥98.0 %
  • Water:
    ≤10 %
  • HPLC:
    ≥97.5 %

Specification Test Results

About this item

Synonyms: Acetic anhydride-[²H₆]
 :
Density:1.14 g/cm³
Flash Pt:54 °C
Storage Temperature:Ambient
EINECS:240-697-0
UN:1715
ADR:8,II
 :GHS07
 :GHS05
 :GHS02
CAS Number:16649-49-3
Formula:C₄D₆O₃
MDL Number:MFCD00044225