Order Entry
Austria
ContactUsLinkComponent
trans-4-(Aminomethyl)cyclohexanecarboxylic acid
trans-4-(Aminomethyl)cyclohexanecarboxylic acid
Catalog # PROOMM0330.00
CAS Number:  
trans-4-(Aminomethyl)cyclohexanecarboxylic acid
Catalog # PROOMM0330.00
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :157.21 g/mol
EINECS:214-818-2
CAS Number:1197-18-8
Formula:C₈H₁₅NO₂
MDL Number:MFCD00001466