Express Eingabe
Austria
ContactUsLinkComponent
trans-4-(Aminomethyl)cyclohexancarbonsäure 98+%
Irritant
trans-4-(Aminomethyl)cyclohexancarbonsäure 98+%
Certificates
undefined
Irritant
trans-4-(Aminomethyl)cyclohexancarbonsäure 98+%

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 %
  • pH:
    6.5 to 8.0 (5 % solution)
  • Titration with HClO4:
    98.0 to 102.0 %

Specification Test Results

Related Information

Frequently Bought Together