Order Entry
Austria
ContactUsLinkComponent
Ebselen, Millipore®
Toxic
Health
Environment
Ebselen, Millipore®
Catalog # 324483-5
CAS Number:  
Toxic
Health
Environment
Ebselen, Millipore®
Catalog # 324483-5
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :274.20 g/mol
UN:3283
ADR:6.1,III
 :GHS06
 :GHS08
 :GHS09
CAS Number:60940-34-3
Formula:C₁₃H₉NOSe
MDL Number:MFCD00210937