
Sodium-4-aminosalicylate dihydrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6018-19-5
:MFCD00151044
Formula:C₇H₆NNaO₃·2H₂O
MDL Number:MFCD00064392
:211.14 g/mol (175.12 anhy)
Melting Pt:250 °C
EINECS:205-091-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together