0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
Hygromycin B
Hygromycin B in solution
Hygromycin B in solution
:
PanReac AppliChem
Hygromycin B in solution
:
PanReac AppliChem
:
31282-04-9
:
:
C₂₀H₃₇N₃O₁₃
:
MFCD06795479
:
527.5 g/mol
Frequently Bought Together
VWR® Standard Line Transfer Pipettes
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
: VWR Collection
VWR® PCR Tubes, 0,2 ml
: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection