G418 disulphate in solution
Supplier: PanReac AppliChem
Certificates
About this item
CAS Number:108321-42-2
:
Formula:C₂₀H₄₄N₄O₁₈S₂
MDL Number:MFCD00058314
:692.72 g/mol
Frequently Bought Together
VWR®, Cell culture media, Dulbecco's Modified Eagles Medium (DMEM)
Supplier: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Penicillin/Streptomycin/Glutamine 100X
Supplier: Biowest
VWR® Basic, Shoe Covers
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Cell culture flasks, Falcon®
Supplier: Corning
VWR®, Cell Strainers
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck