Order Entry
Austria
Orders LinkContactUsLinkComponent
Cable for padlocks, Cobra

Cable for padlocks, Cobra

Black
Catalog # 238-0032
Supplier:   Brady
Cable for padlocks, Cobra
Catalog # 238-0032
Supplier:  Brady
Cable for padlocks, Cobra
Cable for padlocks, Cobra

Black

Price
109,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Flexible cable with protective coating for use with a padlock.

Material: Steel wire, PVC coated

Diameter: 8 mm

Length: 200 cm

Specifications
  • Colour:
    Black
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.