ContactUsLinkComponent
Cut resistant gloves, Showa 67NFW
Cut resistant gloves, Showa 67NFW
Catalog # 112-5881
Supplier:  Showa
undefined
Cut resistant gloves, Showa 67NFW
Catalog # 112-5881
Supplier:  Showa
Specifications
  • Glove size:
    10
  • Ambidextrous / hand-specific:
    Hand-specific
  • Disposable / reusable:
    Reusable
  • Colour:
    Yellow
  • Length:
    305 mm
  • Model:
    67NFW
  • EN norm:
    EN 388:2016 (3142X)
  • Cuff style:
    Gauntlet
  • Liner material:
    Cotton
  • Pk:
    12 PAIR
Specifications
About this item
This soft, cotton flannel liner with natural rubber coating and wrinkle finish provides superior cut, abrasion, snag, and puncture resistance.
  • Wrinkle finish provides outstanding wet and dry grip
  • Protects hands from chemicals and abrasion while remaining elastic and breathable
  • Impermeable for working in damp or greasy environments
  • Ideal replacement for leather and ordinary rubber-coated gloves
It is most suitable for mechanical and automotive applications, construction work, shipping and receiving, and wood handling.
Certifications: EN 388:2016 (3142X); Cat. II
Food contact approved