ContactUsLinkComponent
Sentino® Dispenser Pack Refills for Membrane Filter Dispenser
Sentino® Dispenser Pack Refills for Membrane Filter Dispenser
Certificates
undefined
Sentino® Dispenser Pack Refills for Membrane Filter Dispenser
Certificates
Specifications
  • Catalog No:
  • 514-0628
  • PALL68121ME
  • 514-0629
  • 514-0630
  • 514-0631
  • Type:
  • GN-6 Metricel®
  • ME25
  • Supor®
  • Metricel® Black
  • Metricel® Black
  • Pore size:
  • 0,45 µm
  • 0,45 µm
  • 0,20 µm
  • 0,45 µm
  • 0,80 µm
  • Thickness:
  • 152 µm
  • _
  • 145 µm
  • 117 µm
  • 135 µm
  • Water flow rate:
  • ≥65,0 ml/min/cm² (0,7 bar)
  • _
  • ≥19,3 ml/min/cm² (0,7 bar)
  • ≥34,5 ml/min/cm² (0,7 bar)
  • ≥102,8 ml/min/cm² (0,7 bar)
  • Material:
  • Cellulose mixed esters
  • Cellulose mixed esters
  • Polyethersulfone (PES)
  • Polyethersulfone (PES)
  • Polyethersulfone (PES)