Cut resistant gloves, ChainExtra
Supplier: HONEYWELL SAFETY
Certificates
About this item
These gloves, made of a stainless steel chainmail with detachable plastic straps, provide protection against cuts, a common hazard in meat, fish and processing industries. The gloves are also suitable for use in metal industries.
- Detachable and interchangeable plastic straps with antimicrobial agent
- Firm fastening (press studs) for a secure fit
- Without side slit which may allow a knife to enter at wrist level
- Fitted with an identification tag which can be personalised by the wearer
- Exceptional quality of chain mesh welds, reducing the number of repairs, and increasing the lifetime of the glove
Length: 208 to 283 mm (depending on size)
External diameter of ring: 4 mm
Weight: 149 to 239 g (depending on size)
Certifications: EN 1082
Specifications
- Catalog No:
- 112-2900
- 112-2899
- 112-2896
- 112-2895
- 112-2894
- Model:
- ChainExtra
- ChainExtra
- ChainExtra
- ChainExtra
- ChainExtra
- EN norm:
- EN 1082
- EN 1082
- EN 1082
- EN 1082
- EN 1082
Specifications
Frequently Bought Together