ContactUsLinkComponent
Full face mask, X-plore® 6530
Full face mask, X-plore® 6530
  111-1058
 :  Dräger
undefined
Full face mask, X-plore® 6530
  111-1058
 :  Dräger
  • Type:
    6530 EPDM/Triplex with stainless steel frame
  • Size:
    U*
  • Pk:
    1
The X-plore® 6530 model features breathing filters from the extensive Dräger X-plore® Rd40 filter range and so sets itself apart through being suitable for use in a host of applications.
  • Sealing frame with threefold sealing edge offers reliable protection and a seal-ensuring fit for all facial shapes
  • Comfortable five-point harness makes the product quick to put on and prevents pressure points from forming
  • Distortion-free visor with 180° angle guarantees a large field of vision with a wide, panoramic view
  • Visor made from scratch and chemical resistant Triplex with ultra-stable stainless steel frame (type: 6530 EPDM/Triplex)
  • Intelligent ventilation system with separate inner mask ensures the visor does not become fogged and guarantees an unobstructed view
  • Barcode on the inside makes it easy to record and maintain the mask inventory
  • Integrated stainless steel speech membrane facilitates communication
 : EN 136 Class 3 (CE marking), NIOSH (PC), AS/NZS
 : *One size fits all