Snap ring caps for snap ring vials, N 11
Supplier: MACHEREY-NAGEL
Certificates
About this item
Ready assembled snap ring closures made of polyethylene are available for the snap ring vials.
- Available as hard cap version in transparent and further colours or as soft cap in light blue for easy handling
- Large choice of septa materials
- Most frequent version are closures with cross-slit silicone/PTFE septum, which supports easy penetration of blunt HPLC needles
Frequently Bought Together