Caps for sample tubes, tamper evident
Supplier: Simport Scientific
Certificates
About this item
Caps made of PE feature a long skirt and a super fast thread design allowing them to be removed or sealed with a single turn. Caps are designed with flexible sealing lip which provides leakproof seal.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together