Order Entry
Austria
ContactUsLinkComponent
Cleanroom sorbent pillows and socks, EliminatR®
Cleanroom sorbent pillows and socks, EliminatR®
Catalog # CONTELRPIL
Supplier:  CONTEC
undefined
Cleanroom sorbent pillows and socks, EliminatR®
Catalog # CONTELRPIL
Supplier:  CONTEC

Specifications

  • Description:
    Sorbent pillow
  • Seq:
    01
  • W×D×H:
    305×305×25 mm
  • Pk:
    12

Specifications

About this item

Disposable cleanroom sorbent pillows and socks are designed for large spill pickup or as a drip-catching pad under equipment. They consist of 100% polyester knitted fabric with an durable two-ply polyester outer covering. The pillow is designed for large spill pickup and leak control. The acid-resistant sock is ideal for placement around machinery or drums.

  • Excellent sorbency for aqueous spills
  • Superior holding ability for oils and acids

Pillows and socks are free of any cellulosic contaminants, and are recommended for use in ISO Class 6 or higher cleanrooms.

Certifications: Compatible with gamma, ETO and autoclave sterilization.