ContactUsLinkComponent
Repetitive pipette, Stepper™ 416
Catalog # SOCO416.5000
Supplier:  Socorex Isba
Repetitive pipette, Stepper™ 416
Catalog # SOCO416.5000
Supplier:  Socorex Isba
Specifications
  • Capacity:
    10 - 5000 µl
  • Description:
    Adjustable repetitive pipette, Stepper™ 416
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Adjustable pipette intended for reliable dispensing and high performance results. Selected materials offer outstanding shock resistance and compact shape greatly limit internal contamination. The pipette provides up to 73 doses within a large selection of 53 different volumes.
  • Volumes range from 10 to 5000 µl
  • Unique trigger action (Activation with 4-finger instead of thumb) eliminates thumb fatigue
  • Setting knobs bear clear indication of volumes and number of aliquots
  • Self-locking mechanism
  • Only three positive displacement syringes, colour coded
Syringes are available non sterile (supplied in bulk packages) or sterile (bioproof™ version, individually wrapped). Selection packs include 20 syringes of each volume.
Ordering information: Supplied with colour coded selector buttons, adapter, Ecostep™ syringes.
Syringes
Product Information