Order Entry
Austria
Orders LinkContactUsLinkComponent
Electronic cell counters

Electronic cell counters

Electronic cell counters
Certificates
Electronic cell counters
Electronic cell counters

2 Options Available Below

Product Details & Documents

The electronic memory devices specify blood counting.

  • Power supply operation
  • Rechargeable battery operation
  • Software (download) with computer analysis
  • Big TFT color display
  • Storage of last counting on device

The microscope image can be observed permanently, the numbers can be typed in easily.

Colour: Blue

Delivery information: Every device comes with an instruction manual.

Product Family Options

Product Information

  • Model
    Array
    Availability
    Price
  • 233-5394
    Electronic cell counter
    Electronic cell counter
    AC-10
    10
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 659,00 €
    659,00 €
    1 * 1 ST
       659,00 €

  • HECH40345015
    Electronic cell counter
    Electronic cell counter
    AC-15
    15
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 788,00 €
    788,00 €
    1 * 1 ST
       788,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.