Test kits and reagents for luminometer, BioFix® Lumi
BioFix® products for the determination of biotoxicity.
- In contrast to chemical single parameter analysis, luminescent bacteria tests allow the biotoxicity of a sample to be assessed.
The evaluation is carried out using a luminometer or by determining the oxygen consumption.
Hazardous material: No
- Platform:Microbiological tests
- Storage temperature:−20 ±2 °C
- Shelf life (months):24
- Weight:408 g (Including packaging)
Reagents
Frequently Bought Together