HPTLC plates, silica gel 60, unmodified layers, Nano-ADAMANT
Supplier: MACHEREY-NAGEL
Certificates
About this item
Unmodified HPTLC silica layers available as glass plates with or without fluorescent indicator. The narrow fractionation of the silica particles features theoretical plate heights, which are one order of magnitude smaller than on standard silica layers. Thus, Nano-ADAMANT has the advantage of sharper separations, shorter developing times, and increased detection sensitivity with equal selectivity.
- Good hardness and abrasion resistance due to an optimized binder system
- Increased separation efficiency due to an optimised particle size distribution
- High suitability for trace analyses resulting from a UV indicator with increased brilliance and a low-noise background of the layer
Nano silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 2 to 10 µm.
Frequently Bought Together