Standard conical ground joints cones, long tip, Quickfit®
PYREX® borosilicate glass.
- Useful for construction of stirrer guides
- Avoids the problem of the badge charring at high temperatures which gives an untidy appearance
- Long tip cones manufactured with a minimum shank
Unbadged ground glass cones are free from ceramic markings.
Total length: 90 mm
Frequently Bought Together