Tin standard solution, 1000 mg/L Sn in nitric acid with hydrofluoric acid (each max. 1%) standard for ICP
Supplier: CPACHEM
Certificates
Tin standard solution, 1000 mg/L Sn in nitric acid with hydrofluoric acid (each max. 1%) standard for ICP
Supplier: CPACHEM
About this item
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Sterilised filter bags, Whirl-Pak®
Supplier: NASCO FORT ATKINSON
Nickel standard solution, 10000 mg/L Ni in 5% nitric acid (from Ni), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Turbidity standards, Stablcal®
Supplier: Hach
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
Tin standard solution, 10000 mg/L Sn in 20% hydrochloric acid (from Sn), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
pH buffer solutions, colour coded
Supplier: REAGECON
Buffer Solution pH 7.00 (20°C) (yellow colour) 1 * 1.000 mL
Supplier: PanReac AppliChem
Phenylephrine Hydrochloride, Reference Standard
Supplier: USP