Iron standard solution, 10000 mg/L Fe in 5% hydrochloric acid standard for ICP
Supplier: CPACHEM
Certificates
About this item
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Water Reag. Ph. Eur. 1095506 nitrate-free
Supplier: VWR Chemicals
pH buffer solutions
Supplier: Mettler - Toledo
VWR®, Silicone Rotational viscosity standards
Supplier: VWR Chemicals
Nitrogen (total) CRM, Standard Solutions, Supelco®
Supplier: Merck
Buffer solutions in sachets, perpHect® Pouches, Orion™
Supplier: Thermo Orion
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
Supelco® Organic Reference Standard, Benzotriazole
Supplier: Merck
pH buffer solutions, colour coded
Supplier: Hach