Potassium standard solution, 1,000 mg/L K in water standard for ion chromatography
Supplier: CPACHEM
Certificates
Potassium standard solution, 1,000 mg/L K in water standard for ion chromatography
Supplier: CPACHEM
About this item
Frequently Bought Together
Polyvinyl alcohol, M.W. 22,000 ≥98%, hydrolysed
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW