Niobium standard solution, 1000 mg/L Nb in 5% nitric acid with hydrofluoric acid (max. 1%) standard for ICP
: CPACHEM
Niobium standard solution, 1000 mg/L Nb in 5% nitric acid with hydrofluoric acid (max. 1%) standard for ICP
: CPACHEM
Frequently Bought Together
pH buffers, ready to use
: WTW
pH buffer solutions
: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
pH buffers, DuraCal
: HAMILTON BONADUZ