Iron standard solution, 10000 mg/L Fe in 5% nitric acid standard for ICP
Supplier: CPACHEM
Certificates
About this item
Frequently Bought Together
Isocyanates singles
Supplier: Restek
Indium standard solution, 10 mg/L In in 5% nitric acid (from In), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
PAH-Mix 9 deuterated 100 ng/µl 1 * 1 mL
Supplier: EHRENSTORFER
IBUPROFEN FOR PEAK IDENTIFICATION 1 * 0,09 mg
Supplier: EDQM
Fenchlorazol-ethyl certified 1 * 0,1 g
Supplier: EHRENSTORFER
Oxcarbazepine Related Compounds, Reference Standard
Supplier: USP
Nitra/X/Reagent A 1 * 1 items
Supplier: Lovibond
2,3,4,6-Tetrachlorophenol analytical standard, Supelco®
Supplier: Merck
n-Octacosane, Supelco®
Supplier: Merck
VWR®, Density Standards for the Petroleum Industry, Certified Reference Materials
Supplier: VWR Chemicals
Niobium standard solution, for ICP, in 5% HF, VHG
Supplier: LGC Standards PROMOCHEM
VWR®, Sulphur, Certified Reference Materials, in synthetic diesel
Supplier: VWR Chemicals
Chymotrypsin, Reference Standard
Supplier: USP
Eletriptan Related Compounds, Reference Standard
Supplier: USP
Tetrabromobisphenol A
Supplier: EHRENSTORFER