Order Entry
Austria
ContactUsLinkComponent
 
Boron standard solution, 100 mg/L B in water standard for ICP-MS
Catalog # ACSDM305.W.L1
Supplier:  CPACHEM
CAS Number:  
Boron standard solution, 100 mg/L B in water standard for ICP-MS
Catalog # ACSDM305.W.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Boron
  • Symbol:
    B

Specifications

About this item