Order Entry
Austria
ContactUsLinkComponent
Tetrahydrofuran ≥99.8%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Flammable
Health
Irritant
Tetrahydrofuran ≥99.8%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # 2858-25
CAS Number:  
undefined
Flammable
Health
Irritant
Tetrahydrofuran ≥99.8%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # 2858-25
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    ≥ 99.8 %
  • Assay (C₄H₈O) (by GC, corrected for water):
    ≥99.8 %
  • Peroxide (as H₂O₂):
    ≤0.015 %
  • Residue after Evaporation:
    ≤5 ppm
  • Water (H₂O)(by KF, coulometric):
    ≤0.03 %
  • Optical Absorbance (1-cm path vs water) - 212 nm:
    ≤1.00
  • Optical Absorbance (1-cm path vs water) - 225 nm:
    ≤0.50
  • Optical Absorbance (1-cm path vs water) - 250 nm:
    ≤0.17
  • Optical Absorbance (1-cm path vs water) - 300 nm:
    ≤0.01

Specification Test Results

About this item

 :
 :72.11 g/mol
Merck Index:12,09351
Boiling Pt:65 °C (101.325 kPa)
Melting Pt:–108.3 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:–14.00 °C
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
ADR:3,II
REACH:05-2114615188-48
 :GHS02
 :GHS08
 :GHS07
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356