Order Entry
Austria
ContactUsLinkComponent
Silicon dioxide ≥99.995% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Silicon dioxide ≥99.995% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41096.KS
CAS Number:  
Silicon dioxide ≥99.995% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41096.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7631-86-9
 :
Formula:SiO₂
MDL Number:MFCD00011232
 :60.08 g/mol
Merck Index:14,08493
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1719 °C
Density:2.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-545-4
 :14808-60-7