

Dichloromethane, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 7316.2500
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (by GC):min. 99 %
- Boiling Range:38 - 41 °C
- Preservative (as Amylene):about 20 ppm
- Residue after Evaporation:max. 0.005 %
- Water (H2O):max. 0.1 %
Specification Test Results
About this item
:
:84.93 g/mol
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
ADR:6.1,III
:GHS08
:GHS07
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881